accessibility menu, dialog, popup

UserName

CAS RN 374063-94-2

NNHNOOH

CAS RN 374063-94-2

IUPAC Name: 2-(piperazin-1-yl)pyridine-3-carboxylic acid
Synonyms: 2-(piperazin-1-yl)pyridine-3-carboxylic acid
Molecular Weight (g/mol): 207.23
Molecular Formula: C10H13N3O2
InChi Key: BCDHEUBISMKPOQ-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(N=CC=C1)N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results