accessibility menu, dialog, popup

UserName

CAS RN 373608-42-5

NNHHClHCl

CAS RN 373608-42-5

IUPAC Name: 1-(cyclopropylmethyl)piperazine dihydrochloride
Synonyms: 1-(cyclopropylmethyl)piperazine dihydrochloride
Molecular Weight (g/mol): 213.15
Molecular Formula: C8H18Cl2N2
InChi Key: XOERBUIHFQNXBA-UHFFFAOYSA-N
SMILES: Cl.Cl.C(C1CC1)N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results