CAS RN 3734-48-3
CAS RN 3734-48-3
IUPAC Name:
1,7,8,9,10,10-hexachlorotricyclo[5.2.1.0²,⁶]deca-3,8-diene
Synonyms:
chlordene gamma-chlordene
Molecular Weight (g/mol):
338.86
Molecular Formula:
C10H6Cl6
InChi Key:
XCJXQCUJXDUNDN-UHFFFAOYNA-N
SMILES:
ClC1=C(Cl)C2(Cl)C3C=CCC3C1(Cl)C2(Cl)Cl
1
–
2
of
2
results
1
Chlordene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Chlordene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.