accessibility menu, dialog, popup

UserName

FBFF

CAS RN 373-57-9

IUPAC Name: boron(3+) methanol trifluoride
Synonyms: boron(3+) methanol trifluoride
Molecular Weight (g/mol): 99.85
Molecular Formula: CH4BF3O
InChi Key: VIAZSEYHFKDLSH-UHFFFAOYSA-K
SMILES: [B+3].[F-].[F-].[F-].CO
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)