accessibility menu, dialog, popup

UserName

CAS RN 372-39-4

NH2FF

CAS RN 372-39-4

IUPAC Name: 3,5-difluoroaniline
Synonyms: 3,5-difluoroaniline
Molecular Weight (g/mol): 129.11
Molecular Formula: C6H5F2N
InChi Key: KQOIBXZRCYFZSO-UHFFFAOYSA-N
SMILES: NC1=CC(F)=CC(F)=C1
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)