CAS RN 37148-48-4
CAS RN 37148-48-4
IUPAC Name:
1-(4-amino-3,5-dichlorophenyl)ethan-1-one
Synonyms:
1-(4-amino-3,5-dichlorophenyl)ethanone
Molecular Weight (g/mol):
204.05
Molecular Formula:
C8H7Cl2NO
InChi Key:
JLPKZJDZXIKSCP-UHFFFAOYSA-N
SMILES:
CC(=O)C1=CC(Cl)=C(N)C(Cl)=C1
1
–
2
of
2
results
1
1-(4-Amino-3,5-dichlorophenyl)-2-ethanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-(4-Amino-3,5-dichlorophenyl)-2-ethanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.