accessibility menu, dialog, popup

UserName

CAS RN 37132-68-6

HHClClH2NNHH2N

CAS RN 37132-68-6

IUPAC Name: dihydrogen 3-aminobenzene-1-carboximidamide dichloride
Synonyms: dihydrogen 3-aminobenzenecarboximidamide dichloride
Molecular Weight (g/mol): 208.09
Molecular Formula: C7H11Cl2N3
InChi Key: FKFMMMVFFYPIEE-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NC(=N)C1=CC=CC(N)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results