accessibility menu, dialog, popup

UserName

CAS RN 3710-30-3

H2CCH2

CAS RN 3710-30-3

IUPAC Name: octa-1,7-diene
Synonyms: 1,7-octadiene
Molecular Weight (g/mol): 110.2
Molecular Formula: C8H14
InChi Key: XWJBRBSPAODJER-UHFFFAOYSA-N
SMILES: C=CCCCCC=C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)