CAS RN 370-86-5
CAS RN 370-86-5
IUPAC Name:
1-cyano-N-[4-(trifluoromethoxy)phenyl]methanecarbohydrazonoyl cyanide
Synonyms:
1-cyano-N-[4-(trifluoromethoxy)phenyl]methanecarbohydrazonoyl cyanide
Molecular Weight (g/mol):
254.17
Molecular Formula:
C10H5F3N4O
InChi Key:
BMZRVOVNUMQTIN-UHFFFAOYSA-N
SMILES:
FC(F)(F)OC1=CC=C(NN=C(C#N)C#N)C=C1