CAS RN 36969-89-8
CAS RN 36969-89-8
IUPAC Name:
dimethyl (2-oxoheptyl)phosphonate
Synonyms:
dimethyl 2-oxoheptylphosphonate
Molecular Weight (g/mol):
222.22
Molecular Formula:
C9H19O4P
InChi Key:
LQZCYXCHWNQBKX-UHFFFAOYSA-N
SMILES:
CCCCCC(=O)CP(=O)(OC)OC
1
–
3
of
3
results
1
Dimethyl (2-Oxoheptyl)phosphonate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Dimethyl (2-Oxoheptyl)phosphonate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Dimethyl (2-Oxoheptyl)phosphonate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.