CAS RN 36951-72-1
CAS RN 36951-72-1
IUPAC Name:
1-methyl-4-[7,12,17-tris(1-methylpyridin-1-ium-4-yl)-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1,3,5,7,9,11(23),12,14,16(22),17,19-undecaen-2-yl]pyridin-1-ium tetrakis(4-methylbenzene-1-sulfonate)
Synonyms:
1-methyl-4-[7,12,17-tris(1-methylpyridin-1-ium-4-yl)-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1,3,5,7,9,11(23),12,14,16(22),17,19-undecaen-2-yl]pyridin-1-ium tetratosylate
Molecular Weight (g/mol):
1363.6
Molecular Formula:
C72H66N8O12S4
InChi Key:
AKZFRMNXBLFDNN-UHFFFAOYSA-K
SMILES:
CC1=CC=C(C=C1)S([O-])(=O)=O.CC1=CC=C(C=C1)S([O-])(=O)=O.CC1=CC=C(C=C1)S([O-])(=O)=O.CC1=CC=C(C=C1)S([O-])(=O)=O.C[N+]1=CC=C(C=C1)C1=C2NC(C=C2)=C(C2=NC(C=C2)=C(C2=NC(C=C2)=C(C2=CC=C1N2)C1=CC=[N+](C)C=C1)C1=CC=[N+](C)C=C1)C1=CC=[N+](C)C=C1