CAS RN 36945-98-9
CAS RN 36945-98-9
IUPAC Name:
3-(2-hydroxyphenyl)-6-(3-nitrophenyl)-1,2,3,4-tetrahydropyrimidin-2-one
Synonyms:
icilin
Molecular Weight (g/mol):
311.3
Molecular Formula:
C16H13N3O4
InChi Key:
RCEFMOGVOYEGJN-UHFFFAOYSA-N
SMILES:
OC1=CC=CC=C1N1CC=C(NC1=O)C1=CC=CC(=C1)[N+]([O-])=O
1
–
3
of
3
results
1
Icilin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Icilin, TRC
CAS: 36945-98-9 Molecular Formula: C16H13N3O4 Molecular Weight (g/mol): 311.29 Synonym: 3,6-Dihydro-1-(2-hydroxyphenyl)-4-(3-nitrophenyl)-2(1H)-pyrimidinone,1-(2-Hydroxyphenyl)-4-(3-nitrophenyl)-1,2,3,6-tetrahydropyrimidin-2-one,AG 3-5 IUPAC Name: 3-(2-hydroxyphenyl)-6-(3-nitrophenyl)-1,4-dihydropyrimidin-2-one SMILES: Oc1ccccc1N2CC=C(NC2=O)c3cccc(c3)[N+](=O)[O-]
3