accessibility menu, dialog, popup

UserName

CAS RN 3694-45-9

ClNCS

CAS RN 3694-45-9

IUPAC Name: 1-chloro-4-(isothiocyanatomethyl)benzene
Synonyms: 1-chloro-4-(isothiocyanatomethyl)benzene
Molecular Weight (g/mol): 183.65
Molecular Formula: C8H6ClNS
InChi Key: DEHXIHUIYSXZNH-UHFFFAOYSA-N
SMILES: ClC1=CC=C(CN=C=S)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results