CAS RN 36930-63-9
CAS RN 36930-63-9
IUPAC Name:
5-{[2-(2-iodoacetamido)ethyl]amino}naphthalene-1-sulfonic acid
Synonyms:
iaedans 1,5-I-aedans I-aed
Molecular Weight (g/mol):
434.25
Molecular Formula:
C14H15IN2O4S
InChi Key:
ZMERMCRYYFRELX-UHFFFAOYSA-N
SMILES:
OS(=O)(=O)C1=C2C=CC=C(NCCNC(=O)CI)C2=CC=C1
1
–
3
of
3
results
1
N-(Iodoacetylaminoethyl)-5-Naphthylamine-1-Sulfonic Acid (~90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-(Iodoacetylaminoethyl)-5-Naphthylamine-1-Sulfonic Acid (~90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-(Iodoacetylaminoethyl)-5-Naphthylamine-1-Sulfonic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.