CAS RN 36923-17-8
CAS RN 36923-17-8
IUPAC Name:
7-amino-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
Synonyms:
7-amino-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
Molecular Weight (g/mol):
200.21
Molecular Formula:
C7H8N2O3S
InChi Key:
RJFPBECTFIUTHB-UHFFFAOYNA-N
SMILES:
NC1C2SCC=C(N2C1=O)C(O)=O
1
–
3
of
3
results
1
7-Amino-3-cephem-4-carboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
7-Amino-3-cephem-4-carboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
7-Anca, Thermo Scientific Chemicals
CAS: 36923-17-8 Molecular Formula: C7H8N2O3S Molecular Weight (g/mol): 200.21 InChI Key: RJFPBECTFIUTHB-UHFFFAOYNA-N IUPAC Name: 7-amino-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid SMILES: NC1C2SCC=C(N2C1=O)C(O)=O