accessibility menu, dialog, popup

UserName

CAS RN 369-54-0

FFFF

CAS RN 369-54-0

IUPAC Name: 1,4-bis(difluoromethyl)benzene
Synonyms: 1,4-bis(difluoromethyl)benzene
Molecular Weight (g/mol): 178.13
Molecular Formula: C8H6F4
InChi Key: VWKMZVCSRVFUGW-UHFFFAOYSA-N
SMILES: FC(F)C1=CC=C(C=C1)C(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)