accessibility menu, dialog, popup

UserName

CAS RN 36823-88-8

FFFOClO

CAS RN 36823-88-8

IUPAC Name: 4-(trifluoromethoxy)benzoyl chloride
Synonyms: 4-(trifluoromethoxy)benzoyl chloride
Molecular Weight (g/mol): 224.56
Molecular Formula: C8H4ClF3O2
InChi Key: ZXKKOFJYPRJFIE-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=C(C=C1)C(Cl)=O
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)