accessibility menu, dialog, popup

UserName

CAS RN 368-43-4

FSOO

CAS RN 368-43-4

IUPAC Name: benzenesulfonyl fluoride
Synonyms: benzenesulfonyl fluoride
Molecular Weight (g/mol): 160.16
Molecular Formula: C6H5FO2S
InChi Key: IDIPWEYIBKUDNY-UHFFFAOYSA-N
SMILES: FS(=O)(=O)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results