accessibility menu, dialog, popup

UserName

CAS RN 3678-62-4

CH3NCl

CAS RN 3678-62-4

IUPAC Name: 2-chloro-4-methylpyridine
Synonyms: pyridine, 2-chloro-4-methyl-
Molecular Weight (g/mol): 127.57
Molecular Formula: C6H6ClN
InChi Key: MZVSTDHRRYQFGI-UHFFFAOYSA-N
SMILES: CC1=CC=NC(Cl)=C1
Boiling Point 
  • (5)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (5)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)