CAS RN 363600-92-4
CAS RN 363600-92-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
TLR2-IN-C29, TRC
CAS: 363600-92-4 Molecular Formula: C13 H19 N O3 . H2 S O4 Molecular Weight (g/mol): 335.373 Synonym: TLR2-IN-C29,TLR2-IN-C29,3-[[(2-Hydroxy-3-methoxyphenyl)methylene]amino]-2-methylbenzoic Acid SMILES: CC(C)(C)N1CC(O)c2cc(O)cc(O)c2C1.OS(=O)(=O)O