accessibility menu, dialog, popup

UserName

BrOF

CAS RN 360575-28-6

IUPAC Name: 2-bromo-6-fluorobenzaldehyde
Synonyms: 2-bromo-6-fluorobenzaldehyde
Molecular Weight (g/mol): 203.01
Molecular Formula: C7H4BrFO
InChi Key: PJNILWKRAKKEQM-UHFFFAOYSA-N
SMILES: FC1=C(C=O)C(Br)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)