CAS RN 35999-20-3
IUPAC Name:
3-chloro-1-(4-methoxyphenyl)propan-1-one
Synonyms:
3-chloro-1-(4-methoxyphenyl)propan-1-one
Molecular Weight (g/mol):
198.65
Molecular Formula:
C10H11ClO2
InChi Key:
FJBUZSAECLLZOL-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=C1)C(=O)CCCl
1
–
3
of
3
results
1
3-Chloro-1-(4-methoxyphenyl)propan-1-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3-Chloro-1-(4-methoxyphenyl)propan-1-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3-Chloro-1-(4-methoxyphenyl)propan-1-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.