CAS RN 359861-00-0
CAS RN 359861-00-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
1-[2-O-(Phenoxythioxomethyl)-3,5-O-[1,1,3,3-tetrakis(1-methylethyl)-1,3-disiloxanediyl]-b-D-ribofuranosyl]-1H-1,2,4-triazole-3-carboxamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-[2-O-(Phenoxythioxomethyl)-3,5-O-[1,1,3,3-tetrakis(1-methylethyl)-1,3-disiloxanediyl]-Beta-D-ribofuranosyl]-1H-1,2,4-triazole-3-carboxamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1-[2-O-(Phenoxythioxomethyl)-3,5-O-[1,1,3,3-tetrakis(1-methylethyl)-1,3-disiloxanediyl]-Beta-D-ribofuranosyl]-1H-1,2,4-triazole-3-carboxamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.