accessibility menu, dialog, popup

UserName

CAS RN 35905-85-2

HOONBr

CAS RN 35905-85-2

IUPAC Name: 2-bromopyridine-3-carboxylic acid
Synonyms: 2-bromopyridine-3-carboxylic acid
Molecular Weight (g/mol): 202.01
Molecular Formula: C6H4BrNO2
InChi Key: MMWNKXIFVYQOTK-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CN=C1Br
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)