accessibility menu, dialog, popup

UserName

CAS RN 35836-73-8

H3CH3C(S)OH(R)

CAS RN 35836-73-8

IUPAC Name: 2-[(1R,5S)-6,6-dimethylbicyclo[3.1.1]hept-2-en-2-yl]ethan-1-ol
Synonyms: 2-[(1R,5S)-6,6-dimethylbicyclo[3.1.1]hept-2-en-2-yl]ethanol
Molecular Weight (g/mol): 166.26
Molecular Formula: C11H18O
InChi Key: ROKSAUSPJGWCSM-UWVGGRQHSA-N
SMILES: CC1(C)[C@@H]2C[C@H]1C(CCO)=CC2
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results