accessibility menu, dialog, popup

UserName

CAS RN 35794-11-7

CH3(S)NH2(R)H3C

CAS RN 35794-11-7

IUPAC Name: (3R,5S)-3,5-dimethylpiperidin-1-ium
Synonyms: 3,5-dimethylpiperidine
Molecular Weight (g/mol): 114.21
Molecular Formula: C7H16N
InChi Key: IDWRJRPUIXRFRX-KNVOCYPGSA-O
SMILES: C[C@H]1C[NH2+]C[C@@H](C)C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)