CAS RN 3572-64-3
CAS RN 3572-64-3
IUPAC Name:
2-(3-oxo-2-pentylcyclopentyl)acetic acid
Synonyms:
(3-oxo-2-pentylcyclopentyl)acetic acid
Molecular Weight (g/mol):
212.29
Molecular Formula:
C12H20O3
InChi Key:
PQEYTAGBXNEUQL-UHFFFAOYNA-N
SMILES:
CCCCCC1C(CC(O)=O)CCC1=O
1
–
2
of
2
results
1
(±)-9,10-Dihydrojasmonic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(±)-9,10-Dihydrojasmonic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.