accessibility menu, dialog, popup

UserName

H3CNCH3FFFCl

CAS RN 357-83-5

IUPAC Name: (2-chloro-1,1,2-trifluoroethyl)diethylamine
Synonyms: (2-chloro-1,1,2-trifluoroethyl)diethylamine
Molecular Weight (g/mol): 189.61
Molecular Formula: C6H11ClF3N
InChi Key: BDZHKUAKSMWSAJ-UHFFFAOYNA-N
SMILES: CCN(CC)C(F)(F)C(F)Cl
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results