accessibility menu, dialog, popup

UserName

OHONOOI

CAS RN 35674-27-2

IUPAC Name: 4-iodo-3-nitrobenzoic acid
Synonyms: 4-iodo-3-nitrobenzoic acid
Molecular Weight (g/mol): 293.02
Molecular Formula: C7H4INO4
InChi Key: DNMTZLCNLAIKQC-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=C(I)C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results