accessibility menu, dialog, popup

UserName

CH3NHO

CAS RN 3554-74-3

IUPAC Name: 1-methylpiperidin-3-ol
Synonyms: 1-methylpiperidin-3-ol
Molecular Weight (g/mol): 115.18
Molecular Formula: C6H13NO
InChi Key: UKANCZCEGQDKGF-UHFFFAOYNA-N
SMILES: CN1CCCC(O)C1
Boiling Point 
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)