accessibility menu, dialog, popup

UserName

CAS RN 3550-21-8

NCS

CAS RN 3550-21-8

IUPAC Name: [isothiocyanato(phenyl)methyl]benzene
Synonyms: diphenylmethylisothiocyanate
Molecular Weight (g/mol): 225.31
Molecular Formula: C14H11NS
InChi Key: WDOSFTZMBFYTED-UHFFFAOYSA-N
SMILES: S=C=NC(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)