accessibility menu, dialog, popup

UserName

CAS RN 35120-18-4

CH3OOBr

CAS RN 35120-18-4

IUPAC Name: ethyl 1-bromocyclobutane-1-carboxylate
Synonyms: ethyl 1-bromocyclobutane-1-carboxylate
Molecular Weight (g/mol): 207.07
Molecular Formula: C7H11BrO2
InChi Key: UTVNSHXHFRIXMM-UHFFFAOYSA-N
SMILES: CCOC(=O)C1(Br)CCC1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results