CAS RN 351-52-0
CAS RN 351-52-0
IUPAC Name:
4-(chloromethyl)-2-fluoro-1-methoxybenzene
Synonyms:
4-(chloromethyl)-2-fluoro-1-methoxybenzene
Molecular Weight (g/mol):
174.6
Molecular Formula:
C8H8ClFO
InChi Key:
DDAXEANMRGIVDY-UHFFFAOYSA-N
SMILES:
COC1=CC=C(CCl)C=C1F
1
–
1
of
1
results
1
3-Fluoro-4-methoxybenzyl chloride, 98%
CAS: 351-52-0 Molecular Formula: C8H8ClFO Molecular Weight (g/mol): 174.60 MDL Number: MFCD04973773 InChI Key: DDAXEANMRGIVDY-UHFFFAOYSA-N Synonym: 3-fluoro-4-methoxybenzyl chloride,4-chloromethyl-2-fluoro-1-methoxybenzene,3-fluoro-4-methoxybenzylchloride,benzene,4-chloromethyl-2-fluoro-1-methoxy,benzene, 4-chloromethyl-2-fluoro-1-methoxy,4-chloromethyl-2-fluorophenyl methyl ether,4-chloromethyl-2-fluoro-1-methoxy-benzene,3-fluor-4-methoxy-benzylchlorid,4-chloromethyl-2-fluoroanisole,3-fluoro-4-methoxy benzyl chloride PubChem CID: 2060903 IUPAC Name: 4-(chloromethyl)-2-fluoro-1-methoxybenzene SMILES: COC1=CC=C(CCl)C=C1F