accessibility menu, dialog, popup

UserName

CH3OOSClH3CNH2

CAS RN 350989-77-4

IUPAC Name: ethyl 2-amino-4-(4-chlorophenyl)-5-methylthiophene-3-carboxylate
Synonyms: ethyl 2-amino-4-(4-chlorophenyl)-5-methylthiophene-3-carboxylate
Molecular Weight (g/mol): 295.78
Molecular Formula: C14H14ClNO2S
InChi Key: PJCFNTKRSCPMFG-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)SC(C)=C1C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results