CAS RN 34783-31-8
CAS RN 34783-31-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Hydroxydehydro Nifedipine Carboxylate, TRC
CAS: 34783-31-8 Molecular Formula: C16 H14 N2 O7 Molecular Weight (g/mol): 346.29 Synonym: 3,5-Pyridinedicarboxylic acid, 2-(hydroxymethyl)-6-methyl-4-(2-nitrophenyl)-, 5-methyl ester,4-(2-Nitrophenyl)-2-hydroxymethyl-5-methoxycarbonyl-6-methylpyridine-3-carboxylic acid,4-(2'-Nitrophenyl)-2-hydroxymethyl-5-methoxycarbonyl-6-methyl-pyridine-3-carboxylic acid IUPAC Name: 2-(hydroxymethyl)-5-methoxycarbonyl-6-methyl-4-(2-nitrophenyl)pyridine-3-carboxylic acid SMILES: COC(=O)c1c(C)nc(CO)c(C(=O)O)c1c2ccccc2[N+](=O)[O-]