CAS RN 3457-48-5
CAS RN 3457-48-5
IUPAC Name:
bis(4-methylphenyl)ethane-1,2-dione
Synonyms:
bis(4-methylphenyl)ethane-1,2-dione
Molecular Weight (g/mol):
238.29
Molecular Formula:
C16H14O2
InChi Key:
BCWCEHMHCDCJAD-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1)C(=O)C(=O)C1=CC=C(C)C=C1
1
–
1
of
1
results
1
4,4'-Dimethylbenzil, 98%
CAS: 3457-48-5 Molecular Formula: C16H14O2 Molecular Weight (g/mol): 238.286 MDL Number: MFCD00008554 InChI Key: BCWCEHMHCDCJAD-UHFFFAOYSA-N Synonym: 4,4'-dimethylbenzil,1,2-di-p-tolylethane-1,2-dione,p-tolil,1,2-bis 4-methylphenyl ethane-1,2-dione,ethanedione, bis 4-methylphenyl,ethandione, bis p-tolyl,bis 4-methylphenyl ethane-1,2-dione,1,2-bis 4-methylphenyl-1,2-ethanedione,di-p-tolylethanedione,4,4/'-dimethylbenzil PubChem CID: 76996 IUPAC Name: 1,2-bis(4-methylphenyl)ethane-1,2-dione SMILES: CC1=CC=C(C=C1)C(=O)C(=O)C2=CC=C(C=C2)C