CAS RN 34486-24-3
IUPAC Name:
6-(trifluoromethyl)pyridin-2-amine
Synonyms:
6-(trifluoromethyl)pyridin-2-amine
Molecular Weight (g/mol):
162.12
Molecular Formula:
C6H5F3N2
InChi Key:
NFYYDQMFURPHCC-UHFFFAOYSA-N
SMILES:
NC1=CC=CC(=N1)C(F)(F)F
1
–
1
of
1
results
1
2-Amino-6-(trifluoromethyl)pyridine, 98+%
CAS: 34486-24-3 Molecular Formula: C6H5F3N2 Molecular Weight (g/mol): 162.115 MDL Number: MFCD03093868 InChI Key: NFYYDQMFURPHCC-UHFFFAOYSA-N Synonym: 2-amino-6-trifluoromethyl pyridine,6-trifluoromethyl pyridin-2-amine,2-amino-6-trifluoromethylpyridine,6-trifluoromethyl-2-pyridinamine,6-trifluoromethyl-2-pyridinylamine,6-trifluoromethyl-2-pyridylamine,6-trifluoromethyl-pyridin-2-ylamine,2-pyridinamine, 6-trifluoromethyl,acmc-209i7w PubChem CID: 821024 IUPAC Name: 6-(trifluoromethyl)pyridin-2-amine SMILES: C1=CC(=NC(=C1)N)C(F)(F)F