accessibility menu, dialog, popup

UserName

CAS RN 34160-40-2

BrON

CAS RN 34160-40-2

IUPAC Name: 6-bromopyridine-2-carbaldehyde
Synonyms: 6-bromopyridine-2-carbaldehyde
Molecular Weight (g/mol): 186.01
Molecular Formula: C6H4BrNO
InChi Key: QWFHFNGMCPMOCD-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(C=O)=N1
Molecular Weight (g/mol) 
  • (5)
  • (5)
Quantity 
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (5)
  • (5)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)