accessibility menu, dialog, popup

UserName

H3CNH3CHNNOO

CAS RN 3414-64-0

IUPAC Name: dimethyl[(5-nitro-1H-indol-3-yl)methyl]amine
Synonyms: dimethyl[(5-nitro-1H-indol-3-yl)methyl]amine
Molecular Weight (g/mol): 219.24
Molecular Formula: C11H13N3O2
InChi Key: SVNVKGIWGOHXPW-UHFFFAOYSA-N
SMILES: CN(C)CC1=CNC2=CC=C(C=C12)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results