accessibility menu, dialog, popup

UserName

ClH3CON(R)HO(S)H3CN(R)CH3H3CH

CAS RN 3413-58-9

IUPAC Name: hydrogen (R)-(6-ethoxyquinolin-4-yl)[(2S,5R)-5-ethyl-2,5-dimethyl-1-azabicyclo[2.2.2]octan-2-yl]methanol chloride
Synonyms: optochin
Molecular Weight (g/mol): 404.98
Molecular Formula: C23H33ClN2O2
InChi Key: UBGKQKTVBOWPKR-BVMFEKKJSA-N
SMILES: [H+].[Cl-].CCOC1=CC=C2N=CC=C([C@@H](O)[C@]3(C)CC4CCN3C[C@]4(C)CC)C2=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results