accessibility menu, dialog, popup

UserName

CAS RN 3400-31-5

CH3HNOCl

CAS RN 3400-31-5

IUPAC Name: 2-chloro-N-methylbenzamide
Synonyms: 2-chloro-N-methylbenzamide
Molecular Weight (g/mol): 169.61
Molecular Formula: C8H8ClNO
InChi Key: ITXNDDPDABTCBO-UHFFFAOYSA-N
SMILES: CNC(=O)C1=CC=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results