CAS RN 340-06-7
CAS RN 340-06-7
IUPAC Name:
2,2,2-trifluoro-1-phenylethan-1-ol
Synonyms:
2,2,2-trifluoro-1-phenylethanol
Molecular Weight (g/mol):
176.14
Molecular Formula:
C8H7F3O
InChi Key:
VNOMEAQPOMDWSR-UHFFFAOYNA-N
SMILES:
OC(C1=CC=CC=C1)C(F)(F)F