accessibility menu, dialog, popup

UserName

CAS RN 33924-48-0

CH3OOClOCH3

CAS RN 33924-48-0

IUPAC Name: methyl 5-chloro-2-methoxybenzoate
Synonyms: methyl 5-chloro-2-methoxybenzoate
Molecular Weight (g/mol): 200.62
Molecular Formula: C9H9ClO3
InChi Key: HPTHYBXMNNGQEF-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Cl)=CC=C1OC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results