accessibility menu, dialog, popup

UserName

H3COONCH3ClCl

CAS RN 338954-50-0

IUPAC Name: ethyl 4,6-dichloro-8-methylquinoline-3-carboxylate
Synonyms: ethyl 4,6-dichloro-8-methylquinoline-3-carboxylate
Molecular Weight (g/mol): 284.14
Molecular Formula: C13H11Cl2NO2
InChi Key: DHCZSIOJQCTCHL-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN=C2C(C)=CC(Cl)=CC2=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results