accessibility menu, dialog, popup

UserName

CAS RN 33893-89-9

NNNNN

CAS RN 33893-89-9

IUPAC Name: 5-(pyridin-2-yl)-1H-1,2,3,4-tetrazol-1-ide
Synonyms: 5-(pyridin-2-yl)-1H-1,2,3,4-tetrazol-1-ide
Molecular Weight (g/mol): 146.13
Molecular Formula: C6H4N5
InChi Key: ZKHJXYLAESJIEI-UHFFFAOYSA-N
SMILES: [N-]1N=NN=C1C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results