accessibility menu, dialog, popup

UserName

CAS RN 337508-56-2

IUPAC Name: 1-(bromomethyl)isoquinoline hydrobromide
Synonyms: 1-(bromomethyl)isoquinoline hydrobromide
Molecular Weight (g/mol): 303
Molecular Formula: C10H9Br2N
InChi Key: RWSWRJTWPXSENX-UHFFFAOYSA-N
SMILES: Br.BrCC1=NC=CC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results