accessibility menu, dialog, popup

UserName

CAS RN 3333-52-6

CH3CH3NCH3CH3N

CAS RN 3333-52-6

IUPAC Name: tetramethylbutanedinitrile
Synonyms: tetramethylsuccinonitrile
Molecular Weight (g/mol): 136.2
Molecular Formula: C8H12N2
InChi Key: ZVQXQPNJHRNGID-UHFFFAOYSA-N
SMILES: CC(C)(C#N)C(C)(C)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results