accessibility menu, dialog, popup

UserName

CAS RN 33252-30-1

ClNN

CAS RN 33252-30-1

IUPAC Name: 2-chloropyridine-4-carbonitrile
Synonyms: 2-chloropyridine-4-carbonitrile
Molecular Weight (g/mol): 138.55
Molecular Formula: C6H3ClN2
InChi Key: QRXBTPFMCTXCRD-UHFFFAOYSA-N
SMILES: ClC1=CC(=CC=N1)C#N
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results