accessibility menu, dialog, popup

UserName

N(R)NNHN(R)O(S)(R)OH2NOHHOHOBr

CAS RN 332359-99-6

IUPAC Name: 2-amino-8-bromo-9-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-3H-purin-6-one dihydrate
Synonyms: 2-amino-8-bromo-9-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one dihydrate
Molecular Weight (g/mol): 398.17
Molecular Formula: C10H16BrN5O7
InChi Key: GQDQASJCTAVAAG-UHFFFAOYNA-N
SMILES: O.O.NC1=NC(=O)C2=C(N1)N(C1OC(CO)C(O)C1O)C(Br)=N2
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results