accessibility menu, dialog, popup

UserName

CAS RN 331440-04-1

H3COOHNOCH3

CAS RN 331440-04-1

IUPAC Name: 4-methoxy-N-[(2-methoxyphenyl)methyl]benzamide
Synonyms: 4-methoxy-N-[(2-methoxyphenyl)methyl]benzamide
Molecular Weight (g/mol): 271.32
Molecular Formula: C16H17NO3
InChi Key: OCHBASPTHVVGMS-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)NCC1=CC=CC=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results